Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHRS12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563389
Description
Immunogen sequence: GCMVNKRELT EDGLEKNFAA NTLGVYILTT GLIPVLEKEH DPRVITVSSG GMLVQKLNTN DLQSERTPFD GTMVYAQNKR QQVVLTERW Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 34%.
Putative oxidoreductase.
Specifications
DHRS12 | |
Polyclonal | |
Unconjugated | |
DHRS12 | |
dehydrogenase/reductase (SDR family) member 12; dehydrogenase/reductase 12; dehydrogenase/reductase SDR family member 12; DHRS12; SDR40C1; short chain dehydrogenase/reductase family 40C, member 1; short-chain dehydrogenase/reductase family 40C member 1; zgc:153679 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79758 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0PJE2 | |
DHRS12 | |
Recombinant protein corresponding to Human DHRS12. Recombinant protein control fragment (Product #RP-103286). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction