Learn More
DHRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153931AP
Catalog No. 50-173-0196
Add to cart
Description
Description
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
Specifications
Specifications
DHRS3
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75911, O88876
DD83.1, DHRS3, RDH17, retSDR1, Rsdr1, SDR1, SDR16C1
Rabbit
IgG
150 μL
-20°C
Primary
20148, 313689, 9249
Immunoprecipitation, Western Blot
Unconjugated
DHRS3
Liquid
Dhrs3
DHRS3 Fusion Protein Ag7589
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat