Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17919220UL

 View more versions of this product

Catalog No. NBP17919220


Add to cart

Description

Description

DHRS3 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

DHRS3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
DHRS3
The immunogen for this antibody is Dhrs3. Peptide sequence PGVSATTVLPFHTSTEMFQGMRVRFPNLFPPLKPETVARRTVDAVQQNQA.
Immunogen affinity purified
RUO
Primary
9249
Western Blot
Western Blot 1:1000
NP_035433
DD83.1, dehydrogenase/reductase (SDR family) member 3, EC 1.1.1, EC 1.1.1.300, RDH17, Retinal short-chain dehydrogenase/reductase 1, RETSDR1, retSDR1short chain dehydrogenase/reductase family 16C, member 1, Rsdr1, SDR1, SDR16C1, short-chain dehydrogenase/reductase 1, short-chain dehydrogenase/reductase 3
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit