Learn More
DHRS4L2 Mouse anti-Human, Clone: OTI8A2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810278
Catalog No. 50-166-9984
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DHRS4L2 encodes a member of the short chain dehydrogenase reductase family. The encoded protein may be an NADPH dependent retinol oxidoreductase. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
DHRS4L2
Monoclonal
Unconjugated
DHRS4L2
Lyophilized
DHRS4L2
Full length human recombit protein of human DHRS4L2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI8A2
PBS with 8% trehalose and no preservative; pH 7.3
Q6PKH6
SDR25C3
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
317749