Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS7 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen DHRS7
Immunogen Synthetic peptides corresponding to Dhrs7 (dehydrogenase/reductase (SDR family) member 7) The peptide sequence was selected from the N terminal of Dhrs7. Peptide sequence MVVWVTGASSGIGEELAFQLSKLGVSLVLSARRAQELERVKRRCLENGNL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16891320 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16891320UL
20ul Each for $149.00
Add to cart
 
NBP168913 View Documents Novus BiologicalsSupplier Diversity Partner
NBP168913
100 ul Each for $329.00
Add to cart
 
Description

Description

DHRS7 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

DHRS7
Rabbit
Polyclonal
Unconjugated
51635
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to Dhrs7 (dehydrogenase/reductase (SDR family) member 7) The peptide sequence was selected from the N terminal of Dhrs7. Peptide sequence MVVWVTGASSGIGEELAFQLSKLGVSLVLSARRAQELERVKRRCLENGNL.
Primary
IgG
dehydrogenase/reductase (SDR family) member 7,2310016E22Rik, dehydrogenase/reductase SDR family member 7, DHRS7A, EC 1.1, retDSR4, Retinal short-chain dehydrogenase/reductase 4, RETSDR4, SDR34C1, short chain dehydrogenase/reductase family 34C, member 1
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit