Learn More
Aviva Systems Biology Corp DHRS7B Antibody - middle region (ARP47167_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRS7B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DHRS7B. Alternate names: CGI-93, DKFZp566O084, MGC8916, SDR32C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59478