Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS7C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548632

Catalog No. PA548632


Add to cart

Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Putative oxidoreductase.
Specifications

Specifications

DHRS7C
Polyclonal
PBS with 0.09% sodium azide
A6NNS2
Dehydrogenase/reductase SDR family member 7C, 11--, DHRS7C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
201140
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DHRS7C
Liquid
DHRS7C
KLH conjugated synthetic peptide between 116-145 amino acids from the Central region of human DHRS7C.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit