Learn More
DHS Systems DRASH™ Fatality Management Shelter
Serves as temporary location in field for identification, processing, storage and viewing of casualties following disaster or emergency
Manufacturer:  Reeves Manufacturing RHPK0300
 View more versions of this product
Catalog No. 19-169-424
  Check Availability
Add to cart
Specifications
Description
  • Has different usable sq. ft. of insulated space that can be deployed or taken down
  • Does not require special tools or deal with loose parts
Description & Specifications
Specifications
2-Light set w/electrical set, 3XB Shelter System , Fabric Air Plenum System, 3.5-Ton Mobile ECU
Tan
DRASH Fatality Management Shelter: Small
250 sq. ft.