Learn More
DHS Systems DRASH™ J Series Shelters
Provide more than 1250 sq. ft. of usable interior space for multiple applications
Manufacturer:  Reeves Manufacturing JA201470
 View more versions of this product
Catalog No. 19-167-696
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyester Fabric
419.1cm
165 in.
51.4 in.
1600 lb.
-50° to 131°F
79.5cm
U.S. military standards
J transport and storage trailer, bladder and blower with duct, ladder and sledge hammer, two 1XBTJ vestibule entranceways with built in screen doors, screen windows and Vinyl window set, face plate, T boot and plenum, light and electrical wiring set, ground stakes and low and high wind lines, end wall support kit, field repair kit and illustrated instruction panel
Tan
130.5cm
Shelter
-45.5° to 55°C
726kg
31.33 in.
51.4 x 31.33 x 165 in. (130.5 x 79.5 x 419.1cm)