Learn More
DHS Systems DRASH™ M Series Shelters
Unique shelter design devoid of loose parts or pieces to assemble or misplace
Manufacturer:  Reeves Manufacturing MA100115
 View more versions of this product
Catalog No. 19-167-645
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Center section, end cap, window, door, flooring, Steel stake pin set, repair kit, push poles and transport bag and plenum
1453.8cm
572.4 in.
Shelter section
-45.5° to 55°C
506kg
Green
1402cm
552 in.
1115 lb.
-50° to 131°F
U.S. military standards