DHS Systems DRASH™ S Series Shelters

Available in various dimension of usable space depending on model and variant

Manufacturer: Reeves Manufacturing 00600T

 View more versions of this product

Catalog No. 19-167-657

  Check Availability
Add to Cart

    Specifications

    Product Type Drash Shelter 6S; Stand alone
    Size (English) 402 sq. ft.
    For Use With (Application) Firefighting and Emergency Response
    Color Tan

    • Can be set up within 15 min. of arriving at location by four to six people
    • Can be taken down quickly
    • Do not require any assembly in field, nor require any special equipment for either setting up or striking
    • Pack down to less than 2 percent of deployed size for easy transport
    • Included Components: Complete shelter, integrated flooring, steel stake set, repair kit, push poles, transport bag

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit