Learn More
DHS Systems DRASH™ XB Shelter Series
Available in various dimension of usable space depending on model and variant
Manufacturer:  Reeves Manufacturing 100200T
 View more versions of this product
Catalog No. 19-167-588
  Check Availability
Add to cart
Specifications
Description
  • Can be set up within 15 min. of arriving at location by four to six people
  • Can be taken down quickly
  • Do not require any assembly in field, nor require any special equipment for either setting up or striking
  • Pack down to less than 2 percent of deployed size for easy transport
  • Fits client vehicle or ISO container
  • Included Components: Complete shelter, integrated flooring, steel stake set, repair kit, push poles, transport bag
Description & Specifications
Specifications
Tan
Drash Shelter 2XB; Stand alone
195 sq. ft.
Firefighting and Emergency Response