Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX15, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300118AT

 View more versions of this product

Catalog No. 50-155-6932


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan

DHX15 is a putative ATP-dependent RNA helicase implicated in pre-mRNA splicing.The protein encoded by this gene is a putative ATP-dependent RNA helicase implicated in pre-mRNA splicing.
Specifications

Specifications

DHX15
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
O43143
ATP-dependent RNA helicase No.46, DBP1, DDX15, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15, DEAD/H box-15, DEAH (Asp-Glu-Ala-His) box helicase 15, DEAH (Asp-Glu-Ala-His) box polypeptide 15, DEAH box protein 15, HRH2, PRP43, PRPF43, PrPp43p, putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15, RNA helicase 2
Rabbit
IgG
20 μl
4° C
Primary
1665
Western Blot
Unconjugated
DHX15
liquid
DHX15
Between 150 and 200
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit