Learn More
DHX30 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Manufacturer:  Abnova Corporation H00022907B01P
Catalog No. 89-032-168
Add to cart
Description
Description
Mouse polyclonal antibody raised against a full-length human DHX30 protein.
  • High specificity
  • High reactivity
Specifications
Specifications
DHX30 purified MaxPab mouse
Unconjugated
In 1x PBS, pH 7.4
Liquid
DHX30
DHX30 (AAH14237.1, 1 a.a. ~ 501 a.a) full-length human protein.
RUO
Primary
22907
Western Blot
Mouse polyclonal antibody raised against a full-length human DHX30 protein.
BC014237
DDX30|FLJ11214|KIAA0890|Ret-CoR
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)