Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHX32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113031
Description
DHX32 Polyclonal Antibody for Western Blot, IHC (P), ELISA
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined.
Specifications
DHX32 | |
Polyclonal | |
Unconjugated | |
DHX32 | |
3110079L04Rik; 4732469F02Rik; AA408140; Ddx32; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32; DEAD/H box 32; DEAD/H helicase-like protein 1; DEAD/H helicase-like protein-1; DEAH (Asp-Glu-Ala-His) box polypeptide 32; DEAH box protein 32; DEAH-box helicase 32 (putative); DHLP1; DHX32; helicase DDX32; huDDX32; MuDDX32; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 | |
Rabbit | |
Protein G | |
RUO | |
101437, 55760 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q7L7V1, Q8BZS9 | |
DHX32 | |
Recombinant Human Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction