Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX37, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300856AM

 View more versions of this product

Catalog No. 50-155-8628


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.
Specifications

Specifications

DHX37
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q8IY37
DDX37, DEAD/DEAH box helicase DDX37, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37, DEAH box protein 37, probable ATP-dependent RNA helicase DHX37
Rabbit
IgG
100 μl
4° C
Primary
57647
Western Blot
Unconjugated
DHX37
liquid
DHX37
Between 1 and 50
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit