Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHX57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557548
Description
Immunogen sequence: KRQFTELLSD IGFAREGLRA REIEKRAQGG DGVLDATGEE ANSNAENPKL ISAMLCAALY PNVVQVKSPE GKFQKTSTGA VR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Probable ATP-binding RNA helicase.
Specifications
DHX57 | |
Polyclonal | |
Unconjugated | |
DHX57 | |
AW494914; DDX57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 homolog; DEAH box protein 57; DEAH-box helicase 57; DEAH-box RNA/DNA helicase AAM73547; DExH-box helicase 57; DHX57; Putative ATP-dependent RNA helicase DHX57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P158 | |
DHX57 | |
Recombinant protein corresponding to Human DHX57. Recombinant protein control fragment (Product #RP-96180). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction