Alfa Aesar™ Di-2-thienyl ketone, 98%

Manufacturer: Alfa Aesar™ A1206214

Quantity 25g

 View more versions of this product

Catalog No. AAA1206214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 704-38-1
    Molecular Formula C9H6OS2
    Formula Weight 194.28
    MDL Number MFCD00052096
    Beilstein 129168
    Synonym Bis-2-thienyl ketone; 2-(2-Thenoyl)thiophene
    Chemical Name or Material Di-2-thienyl ketone

    Specifications

    Formula Weight 194.28
    Molecular Formula C9H6OS2
    Quantity 25g
    MDL Number MFCD00052096
    CAS 704-38-1
    Beilstein 129168
    Melting Point 87° to 90°C
    Synonym Bis-2-thienyl ketone; 2-(2-Thenoyl)thiophene
    Chemical Name or Material Di-2-thienyl ketone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit