Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%, Thermo Scientific Chemicals

Catalog Number AC348980250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
30698-64-7
C12H36Cl2Cu2N4O2
466.44
MFCD01321163
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
11190467
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
30698-64-7
C12H36Cl2Cu2N4O2
466.44
MFCD01321163
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
11190467
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136.0°C to 138.0°C
Color Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 464.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Di-μ-hydroxo-bis-[(N,N,N′,N′-tetramethylethylenediamine)copper(II)]chloride
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit