Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-μ-iodobis(p-cymene)iodoruthenium(II) 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
90614-07-6
C20H28I4Ru2
MFCD08064027
90614-07-6
C20H28I4Ru2
MFCD08064027
Quantity 250mg
Packaging Glass bottle
Color Bordeaux
Infrared Spectrum Authentic
Elemental Analysis 96.0% min.
Assay Percent Range 97%
Formula Weight 978.18
Physical Form Crystalline Powder
Chemical Name or Material Di-μ-iodobis(p-cymene)iodoruthenium(II)

GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.