Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H3K17me2aK18ac Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540094
Description
H3K17me2aK18ac Polyclonal antibody specifically detects H3K17me2aK18ac in Human, Mouse samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Peptide Array, Western Blot.Specifications
H3K17me2aK18ac | |
Polyclonal | |
Unconjugated | |
H3C12 | |
Acetyl H3; Acetyl Histone H3; Di histone H3/c; dimethyl H3; di-methyl H3; Dimethyl Histone H3; di-methyl histone H3; Di-Methyl-Acetyl-H3 Arg17 Lys18; Di-Methyl-Acetyl-H3 R17 K18; Di-Methyl-Acetyl-Histone H3 Arg17 Lys18; Di-Methyl-Acetyl-Histone H3 R17 K18; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K17me2aK18ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3g; H3h; H3i; H3K17me2; H3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Peptide Array, Western Blot | |
0.94 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
P68431, P68433, P84228, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
KLH-conjugated synthetic peptide corresponding to the region of human histone H3 containing the asymmetrically dimethylated R17 and the acetylated lysine 18 (H3R17me2(asym). | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction