Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-mu-chlorobis(norbornadiene)dirhodium(I), Rh 44% min

Click to view available options
Quantity:
0.5g
1g
12257-42-0
C14H16Cl2Rh
358.087
MFCD00198060
HCZNBZKRJTYNCE-UHFFFAOYSA-L
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer,bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride,bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
114600
bicyclo[2.2.1]hepta-2,5-diene;rhodium(2+);dichloride
C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Cl-].[Cl-].[Rh+2]
12257-42-0
C14H16Cl2Rh
358.087
MFCD00198060
HCZNBZKRJTYNCE-UHFFFAOYSA-L
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer,bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride,bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
114600
bicyclo[2.2.1]hepta-2,5-diene;rhodium(2+);dichloride
C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Cl-].[Cl-].[Rh+2]
Odor Odorless
Quantity 1g
Concentration or Composition (by Analyte or Components) Rh 44% min.
Solubility Information Slightly soluble in acetone,chloroform,methanol and insoluble in water.
Formula Weight 461.01
Physical Form Powder
Chemical Name or Material Di-mu-chlorobis(norbornadiene)dirhodium(I)

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-510-4

TSCA : No

Recommended Storage : Ambient temperatures