Alfa Aesar™ Di-n-butyl phosphate, 96%

Manufacturer: Alfa Aesar™ 43371AK

Quantity 250mL

 View more versions of this product

Catalog No. AA43371AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-66-4
    Molecular Formula (CH3(CH2)3O)2P(O)OH
    Formula Weight 210.21
    MDL Number MFCD00015245
    UN Number UN1718
    Chemical Name or Material Di-n-butyl phosphate

    Specifications

    Formula Weight 210.21
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.058g/mL
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point −13°C
    Quantity 250mL
    Flash Point 157°C (315°F)
    Odor Odorless
    pH 1.0
    Refractive Index 1.428
    Molecular Formula (CH3(CH2)3O)2P(O)OH
    MDL Number MFCD00015245
    CAS 107-66-4
    Health Hazard 1 Danger
    RTECS Number TB9605000
    TSCA TSCA
    UN Number UN1718
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYL ACID PHOSPHATE
    Boiling Point 250°C
    Chemical Name or Material Di-n-butyl phosphate
    Assay 96%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit