Alfa Aesar™ Di-n-butyl sulfoxide, 97%

Manufacturer: Alfa Aesar™ B2367214

Quantity 25g

 View more versions of this product

Catalog No. AAB2367214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2168-93-6
    Molecular Formula C8H18OS
    Formula Weight 162.3
    MDL Number MFCD00002093
    Beilstein 1747425
    Synonym n-Butyl sulfoxide
    Chemical Name or Material Di-n-butyl sulfoxide

    Specifications

    Formula Weight 162.3
    Melting Point 31° to 34°C
    Quantity 25g
    Flash Point 121°C (249°F)
    Boiling Point 250°C
    Molecular Formula C8H18OS
    MDL Number MFCD00002093
    CAS 2168-93-6
    Beilstein 1747425
    Synonym n-Butyl sulfoxide
    RTECS Number EK5700000
    Chemical Name or Material Di-n-butyl sulfoxide
    Assay 97%

    Description

    Applications
    It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.

    Solubility
    Slightly soluble in water.

    Notes
    Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit