Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butylamine, for HPLC, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
Packaging:
Glass bottle
111-92-2
C8H19N
129.24
MFCD00009429
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine,di-n-butylamine,1-butanamine, n-butyl,n-butyl-1-butanamine,dibutilamina,n-dibutylamine,di-n-butyl amine,dibutyl amine,di-normal-butylamine,dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC
111-92-2
C8H19N
129.24
MFCD00009429
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine,di-n-butylamine,1-butanamine, n-butyl,n-butyl-1-butanamine,dibutilamina,n-dibutylamine,di-n-butyl amine,dibutyl amine,di-normal-butylamine,dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC
Viscosity 0.9 mPa.s (20°C)
Refractive Index 1.4160 to 1.4180
Transmittance 99 (c=0.01M water, 254nm) min.
Residue after Evaporation 0.0005% max.
Melting Point -62.0°C
Water 0.1% max.
Specific Gravity 0.76
Quantity 100mL
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Packaging Glass bottle
Color White
Linear Formula [CH3(CH2)3]2NH
Merck Index 15, 3037
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Formula Weight 129.24
Physical Form Crystalline Free Flowing Powder
Percent Purity ≥99% (GC)
Grade HPLC
Chemical Name or Material Di-n-butylamine
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre

GHS Signal Word: Danger

EINECSNumber : 203-921-8