Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butylmagnesium, 0.5M solution in heptane, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
1191-47-5,142-82-5,97-93-8
C8H18Mg
138.54
MFCD00015225
KJJBSBKRXUVBMX-UHFFFAOYSA-N
dibutylmagnesium,magnesium, dibutyl,di-n-butylmagnesium,magnesium,butyl 1-methylpropyl,dibutyl magnesium,di-n-butyl magnesium,di-n-butylmagnesium solution,odhfjiddbsdwnu-uhfffaoysa-n,di-n-butylmagnesium, 0,5m in heptane,di-n-butylmagnesium solution, 1.0 m in heptane
70929
magnesium;butane
CCC[CH2-].CCC[CH2-].[Mg+2]
1191-47-5,142-82-5,97-93-8
C8H18Mg
138.54
MFCD00015225
KJJBSBKRXUVBMX-UHFFFAOYSA-N
dibutylmagnesium,magnesium, dibutyl,di-n-butylmagnesium,magnesium,butyl 1-methylpropyl,dibutyl magnesium,di-n-butyl magnesium,di-n-butylmagnesium solution,odhfjiddbsdwnu-uhfffaoysa-n,di-n-butylmagnesium, 0,5m in heptane,di-n-butylmagnesium solution, 1.0 m in heptane
70929
magnesium;butane
CCC[CH2-].CCC[CH2-].[Mg+2]
Viscosity 0.9 mPa.s (22°C)
Melting Point -71.0°C
Appearance Clear, colorless to slightly hazy solution
Flash Point −4°C
Quantity 100mL
Density 0.7300g/mL
Specific Gravity 0.73
Packaging AcroSeal™ Glass bottle
Linear Formula (C4H9)2Mg
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 138.54
Chemical Name or Material Di-n-butylmagnesium
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
In contact with water releases

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 214-736-7