Alfa Aesar™ Di-n-butyltin bis(2,4-pentanedionate), 95%

Manufacturer: Alfa Aesar™ B2392118

Quantity 50g

 View more versions of this product

Catalog No. AAB2392118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22673-19-4
    Molecular Formula C18H32O4Sn
    Formula Weight 431.16
    MDL Number MFCD00077994
    UN Number UN1992
    Chemical Name or Material Di-n-butyltin bis(2,4-pentanedionate)

    Specifications

    Formula Weight 431.16
    Boiling Point 150°C
    TSCA TSCA
    Melting Point 26°C
    Molecular Formula C18H32O4Sn
    MDL Number MFCD00077994
    CAS 22673-19-4
    Quantity 50g
    UN Number UN1992
    Flash Point 40°C (104°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H302-H312-H332-H315-H319-H335
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Weak
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material Di-n-butyltin bis(2,4-pentanedionate)
    Assay 95%

    GHS H Statement
    H226-H302-H312-H332-H315-H319-H335
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit