Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-n-butyltin bis(2,4-pentanedionate), 95%

Click to view available options
Quantity:
250g
1000g
22673-19-4
C18H32O4Sn
MFCD00077994
dibutyltin bis acetylacetonate,di-n-butylbis 2,4-pentanedionate tin,dibutyltin bis 2,4-pentanedionate,3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z,3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one,z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one
22673-19-4
C18H32O4Sn
MFCD00077994
dibutyltin bis acetylacetonate,di-n-butylbis 2,4-pentanedionate tin,dibutyltin bis 2,4-pentanedionate,3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z,3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one,z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one
Flash Point 40°C (104°F)
Melting Point 26°C
Quantity 1000g
Odor Weak
Boiling Point 150°C
UN Number UN1992
Formula Weight 431.16
Percent Purity 95%
Chemical Name or Material Di-n-butyltin bis(2,4-pentanedionate)

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA