Alfa Aesar™ Di-n-butyltin bis(2-ethylhexanoate)

Manufacturer: Alfa Aesar™ 7112622

Quantity 100g

 View more versions of this product

Catalog No. AA7112622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2781-10-4
    Molecular Formula C24H48O4Sn
    Formula Weight 519.34
    MDL Number MFCD00015261
    UN Number UN3146
    Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)

    Specifications

    Physical Form Lumps
    Formula Weight 519.34
    Boiling Point 220°C (2mmHg)
    Molecular Formula C24H48O4Sn
    Quantity 100g
    Solubility Information Soluble in mineral oil, acetone, xylene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H310-H361
    Toxic if swallowed.
    Fatal in contact with skin.
    Suspected of damaging fertility or the unborn child.
    Health Hazard 3 GHS P Statement
    P280h-P273-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Density 1.070g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    RTECS Number WH6714500
    TSCA TSCA
    Melting Point 57° to 59°C
    MDL Number MFCD00015261
    UN Number UN3146
    Odor Odorless
    CAS 2781-10-4
    Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)

    GHS H Statement
    H301-H310-H361
    Toxic if swallowed.
    Fatal in contact with skin.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P280h-P273-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit