Alfa Aesar™ Di-n-butyltin dichloride, 96%

Organotin intermediate

Manufacturer: Alfa Aesar™ 1411618

Quantity 50g

 View more versions of this product

Catalog No. AA1411618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 683-18-1
    Linear Formula [CH3(CH2)3]2SnCl2
    Formula Weight 303.83
    MDL Number MFCD00000518
    UN Number UN2928
    Synonym Di-n-butyldichlorostannane
    Chemical Name or Material Di-n-butyltin dichloride

    Specifications

    Formula Weight 303.83
    Health Hazard 2 GHS H Statement
    H301-H330-H360-H372-H341-H314-H312
    Toxic if swallowed.
    Fatal if inhaled.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Causes severe skin burns and eye damage.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 135°C (10mmHg)
    MDL Number MFCD00000518
    Quantity 50g
    Solubility Information Insoluble in cold water; Hydrolyzed by hot water; Soluble in organic solvents
    Flash Point 168° (334°F)
    Odor Acrid
    RTECS Number WH7100000
    UN Number UN2928
    Synonym Di-n-butyldichlorostannane
    Health Hazard 1 Danger
    Density 1.36g/mL
    Color White
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Sensitivity Moisture Sensitive
    TSCA TSCA
    Melting Point 39° to 41°C
    Physical Form Crystalline
    Linear Formula [CH3(CH2)3]2SnCl2
    CAS 683-18-1
    Refractive Index 1.4991
    Chemical Name or Material Di-n-butyltin dichloride
    Assay 96%

    GHS H Statement
    H301-H330-H360-H372-H341-H314-H312
    Toxic if swallowed.
    Fatal if inhaled.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Causes severe skin burns and eye damage.
    Harmful in contact with skin.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit