Alfa Aesar™ Di-n-butyltin oxide

Manufacturer: Alfa Aesar™ 1411718

Quantity 50g

 View more versions of this product

Catalog No. AA1411718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 818-08-6
    Linear Formula [CH3(CH2)3]2SnO
    Formula Weight 248.92
    MDL Number MFCD00001992
    UN Number UN3146
    Beilstein 4126243
    Synonym Dibutyloxotin
    Chemical Name or Material Di-n-butyltin oxide

    Specifications

    Formula Weight 248.92
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00001992
    Health Hazard 2 GHS H Statement
    H300-H319
    Fatal if swallowed.
    Causes serious eye irritation.
    Odor Odorless
    RTECS Number WH7175000
    Melting Point >300°C
    CAS 818-08-6
    UN Number UN3146
    Synonym Dibutyloxotin
    Health Hazard 1 Danger
    Density 1.58g/mL
    Color White
    TSCA TSCA
    Physical Form Powder
    Linear Formula [CH3(CH2)3]2SnO
    Quantity 50g
    Beilstein 4126243
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Chemical Name or Material Di-n-butyltin oxide

    GHS H Statement
    H300-H319
    Fatal if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit