Alfa Aesar™ Di-n-butyltin sulfide

Manufacturer: Alfa Aesar™ 3961809

Quantity 10g

 View more versions of this product

Catalog No. AA3961809

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4253-22-9
    Formula Weight 264.99
    MDL Number MFCD00015528
    UN Number UN2788
    Chemical Name or Material Di-n-butyltin sulfide

    Specifications

    Formula Weight 264.99
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 4253-22-9
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    TSCA TSCA
    MDL Number MFCD00015528
    Quantity 10g
    UN Number UN2788
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Chemical Name or Material Di-n-butyltin sulfide

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit