Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Di-n-decyl sulfide, 97%
Click to view available options
Quantity:
10g
50g
Specifications
Melting Point | 25°C to 28°C |
Quantity | 10g |
Boiling Point | 217°C to 218°C (8mmHg) |
Beilstein | 1763427 |
Formula Weight | 314.63 |
Percent Purity | 97% |
Chemical Name or Material | Di-n-decyl sulfide |
Safety and Handling
EINECSNumber : 211-761-5
TSCA : Yes
Recommended Storage : Ambient temperatures