Alfa Aesar™ Di-n-dodecyl sulfide, 90+%

Manufacturer: Alfa Aesar™ B2369622

Quantity 100g

 View more versions of this product

Catalog No. AAB2369622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2469-45-6
    Molecular Formula [CH3(CH2)11]2S
    Formula Weight 370.73
    MDL Number MFCD00008975
    Beilstein 1777013
    Synonym n-Dodecyl sulfide
    Chemical Name or Material Di-n-dodecyl sulfide

    Specifications

    Formula Weight 370.73
    Melting Point 38° to 42°C
    Boiling Point 260° to 263°C (4mmHg)
    MDL Number MFCD00008975
    CAS 2469-45-6
    Synonym n-Dodecyl sulfide
    TSCA TSCA
    Quantity 100g
    Molecular Formula [CH3(CH2)11]2S
    Beilstein 1777013
    Chemical Name or Material Di-n-dodecyl sulfide
    Assay ≥90%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit