Alfa Aesar™ Di-n-hexyl sulfide, 97%

Manufacturer: Alfa Aesar™ B2232522

Quantity 100g

 View more versions of this product

Catalog No. AAB2232522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6294-31-1
    Molecular Formula [CH3(CH2)5]2S
    Formula Weight 202.4
    MDL Number MFCD00009528
    UN Number UN3334
    Beilstein 1738333
    Synonym n-Hexyl sulfide
    Chemical Name or Material Di-n-hexyl sulfide

    Specifications

    Formula Weight 202.4
    TSCA TSCA
    Boiling Point 260°C
    Quantity 100g
    Flash Point 116°C (241°F)
    Odor Stench
    Molecular Formula [CH3(CH2)5]2S
    MDL Number MFCD00009528
    CAS 6294-31-1
    UN Number UN3334
    Beilstein 1738333
    Synonym n-Hexyl sulfide
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
    Chemical Name or Material Di-n-hexyl sulfide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit