Alfa Aesar™ Di-n-octadecyl disulfide, 98%

Manufacturer: Alfa Aesar™ B2197803

Quantity 1g

 View more versions of this product

Catalog No. AAB2197803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2500-88-1
    Molecular Formula CH3(CH2)17S-S(CH2)17CH3
    Formula Weight 571.11
    MDL Number MFCD00026687
    Beilstein 1715376
    Synonym n-Octadecyl disulfide
    Chemical Name or Material Di-n-octadecyl disulfide

    Specifications

    Formula Weight 571.11
    TSCA TSCA
    Molecular Formula CH3(CH2)17S-S(CH2)17CH3
    MDL Number MFCD00026687
    CAS 2500-88-1
    Beilstein 1715376
    Synonym n-Octadecyl disulfide
    Odor Unpleasant
    Melting Point 60° to 64°C
    Quantity 1g
    Chemical Name or Material Di-n-octadecyl disulfide
    Assay 98%

    Description

    Applications
    It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit