Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-n-octyldichlorosilane, 97%

Click to view available options
Quantity:
10g
50g
18416-07-4
C16H34Cl2Si
325.433
MFCD01074472
CVAGYCLEIYGJQT-UHFFFAOYSA-N
di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
248429
dichloro(dioctyl)silane
CCCCCCCC[Si](CCCCCCCC)(Cl)Cl
18416-07-4
C16H34Cl2Si
325.433
MFCD01074472
CVAGYCLEIYGJQT-UHFFFAOYSA-N
di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
248429
dichloro(dioctyl)silane
CCCCCCCC[Si](CCCCCCCC)(Cl)Cl
Linear Formula [CH3(CH2)7]2SiCl2
UN Number UN2987
Solubility Information Sparingly Soluble (8.8E-6g/L) at 25°C.
Formula Weight 325.44
Physical Form Liquid
Percent Purity 97%
Density 0.940 g/mL
Sensitivity Moisture sensitive
Boiling Point 145°C (0.2mmHg)
Color Colorless to Yellow
Quantity 10g
Chemical Name or Material Di-n-octyldichlorosilane
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen