Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-n-pentyl phthalate, 98%

Manufacturer:  Alfa Aesar™ B2291022

Quantity 100g

 View more versions of this product

Catalog No. AAB2291022


  Check Availability
Add to cart

Specifications

Description

Applications
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

131-18-0
306.41
UN3082
Di-n-pentyl phthalate
C18H26O4
MFCD00041934
Di-n-pentyl phthalate; Phthalic acid di-n-pentyl ester

Specifications

118°C (244°F)
342°C
131-18-0
306.41
UN3082
Di-n-pentyl phthalate
Odorless
100g
C18H26O4
MFCD00041934
Di-n-pentyl phthalate; Phthalic acid di-n-pentyl ester
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : TI1930000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit