Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-n-pentyl phthalate, 98%

Click to view available options
Quantity:
5g
25g
100g
131-18-0
C18H26O4
306.402
MFCD00041934
IPKKHRVROFYTEK-UHFFFAOYSA-N
dipentyl phthalate,di-n-pentyl phthalate,diamyl phthalate,amyl phthalate,amoil,di-n-amyl phthalate,di-n-pentylphthalate,phthalic acid, dipentyl ester,phthalic acid diamyl ester,1,2-benzenedicarboxylic acid, dipentyl ester
8561
CHEBI:34680
dipentyl benzene-1,2-dicarboxylate
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
131-18-0
C18H26O4
306.402
MFCD00041934
IPKKHRVROFYTEK-UHFFFAOYSA-N
dipentyl phthalate,di-n-pentyl phthalate,diamyl phthalate,amyl phthalate,amoil,di-n-amyl phthalate,di-n-pentylphthalate,phthalic acid, dipentyl ester,phthalic acid diamyl ester,1,2-benzenedicarboxylic acid, dipentyl ester
8561
CHEBI:34680
dipentyl benzene-1,2-dicarboxylate
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
Applications
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Quantity 5g
Odor Odorless
Boiling Point 342°C
Flash Point 118°C (244°F)
UN Number UN3082
Formula Weight 306.41
Percent Purity 98%
Chemical Name or Material Di-n-pentyl phthalate

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : TI1930000

TSCA : TSCA