Alfa Aesar™ Di-n-propyl phthalate, 98%

Manufacturer: Alfa Aesar™ B2405609

Quantity 10g

 View more versions of this product

Catalog No. AAB2405609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 131-16-8
    Molecular Formula C14H18O4
    Formula Weight 250.3
    MDL Number MFCD00009371
    UN Number UN3082
    Beilstein 2332522
    Synonym Benzene-1,2-dicarboxylic acid di-n-propyl ester; Phthalic acid di-n-propyl ester
    Chemical Name or Material Di-n-propyl phthalate

    Specifications

    Formula Weight 250.3
    Flash Point >110°C (230°F)
    Quantity 10g
    TSCA TSCA
    Molecular Formula C14H18O4
    MDL Number MFCD00009371
    CAS 131-16-8
    UN Number UN3082
    Beilstein 2332522
    Synonym Benzene-1,2-dicarboxylic acid di-n-propyl ester; Phthalic acid di-n-propyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    RTECS Number TI1940000
    Boiling Point 168° to 170°C (3mmHg)
    Chemical Name or Material Di-n-propyl phthalate
    Assay 98%

    Description

    Applications
    Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

    Solubility
    Soluble in chloroform, methanol.

    Notes
    Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit