Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-p-toluoyl-D-tartaric acid monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
71607-31-3
C20H18O8·H2O
404.38
FOTRUJUPLHRVNU-FAESNJTISA-N
+-o,o'-di-p-toluoyl-d-tartaric acid monohydrate,2r,3s-2,3-bis 4-methylbenzoyloxy butanedioic acid hydrate,2s,3s-+-2,3-bis 4-methylbenzoyl oxy succinic acid monohydrate
71299557
(2R,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O
71607-31-3
C20H18O8·H2O
404.38
FOTRUJUPLHRVNU-FAESNJTISA-N
+-o,o'-di-p-toluoyl-d-tartaric acid monohydrate,2r,3s-2,3-bis 4-methylbenzoyloxy butanedioic acid hydrate,2s,3s-+-2,3-bis 4-methylbenzoyl oxy succinic acid monohydrate
71299557
(2R,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O
Specific Rotation + 140.00
Infrared Spectrum Authentic
Melting Point 163.0°C to 165.0°C
Quantity 25g
Water 6% max.
Assay Percent Range 97.5% min. (HPLC)
Linear Formula [CH3C6H4CO2CH(CO2H)-]2·H2O
Fieser 01,351
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and acetone
Formula Weight 404.38
Percent Purity 98%
Chemical Name or Material Di-p-toluoyl-D-tartaric acid monohydrate
Show More Show Less