Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Di-p-toluoyl-L-tartaric acid, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
32634-66-5
C20H18O8
386.36
MFCD00064440
FOTRUJUPLHRVNU-QNBGGDODSA-N
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid,di-p-toluoyl-l-tartaric acid,l-dtta,--di-p-toluoyl-l-tartaric acid,--di-1,4-o-toluoyl-l-tartaric acid,--o,o'-di-p-toluoyl-l-tartaric acid,l---di-p-toluoyltartaric acid,2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid,2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid,butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
12297760
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O
32634-66-5
C20H18O8
386.36
MFCD00064440
FOTRUJUPLHRVNU-QNBGGDODSA-N
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid,di-p-toluoyl-l-tartaric acid,l-dtta,--di-p-toluoyl-l-tartaric acid,--di-1,4-o-toluoyl-l-tartaric acid,--o,o'-di-p-toluoyl-l-tartaric acid,l---di-p-toluoyltartaric acid,2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid,2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid,butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
12297760
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O
Specific Rotation -142
Color White to Yellow
Water 0.5% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10g
Melting Point 164°C to 170°C
Specific Rotation Condition −142° (20°C c=10,C2H5OH)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [CH3C6H4CO2CH(CO2H)-]2
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and ethanol
Formula Weight 386.36
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material (-)-Di-p-toluoyl-L-tartaric acid
Show More Show Less