Alfa Aesar™ Di-p-toluoyl-L-tartaric acid, 98%

Manufacturer: Alfa Aesar™ B2297222

Quantity 100g

 View more versions of this product

Catalog No. AAB2297222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32634-66-5
    Molecular Formula C20H18O8
    Formula Weight 386.36
    MDL Number MFCD00064440
    Beilstein 2022481
    Chemical Name or Material Di-p-toluoyl-L-tartaric acid

    Specifications

    Formula Weight 386.36
    Melting Point 166° to 171°C
    Quantity 100g
    Optical Rotation −139° (c=1 in Ethanol)
    TSCA TSCA
    Molecular Formula C20H18O8
    MDL Number MFCD00064440
    CAS 32634-66-5
    Beilstein 2022481
    Chemical Name or Material Di-p-toluoyl-L-tartaric acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit