Alfa Aesar™ Di-p-tolyl ether, 99%

Manufacturer: Alfa Aesar™ H5516714

Quantity 25g

 View more versions of this product

Catalog No. AAH5516714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1579-40-4
    Formula Weight 198.26
    MDL Number MFCD00025979
    UN Number UN3077
    Synonym p-Tolyl ether; 4,4'-Dimethyldiphenyl ether
    Chemical Name or Material Di-p-tolyl ether

    Specifications

    Formula Weight 198.26
    Melting Point 47° to 49°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00025979
    Synonym p-Tolyl ether; 4,4'-Dimethyldiphenyl ether
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Physical Form Solid
    CAS 1579-40-4
    Boiling Point 285°C
    Color Pale yellow
    Quantity 25g
    UN Number UN3077
    RTECS Number DA6625000
    Chemical Name or Material Di-p-tolyl ether
    Assay 99%

    Description

    Applications
    Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.

    Solubility
    Soluble in acetone, benzene, ether.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit