Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Di-sec-butoxyaluminoxytriethoxysilane

Click to view available options
Quantity:
10g
50g
250g
68959-06-8
C14H33AlO6Si
352.479
MFCD00053771
MTHHTRVJDXYJHE-UHFFFAOYSA-N
di-sec-butoxyaluminoxytriethoxysilane,bis butan-2-olato triethyl orthosilicato-o' aluminium,aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o',aluminum, bis 2-butanolato triethyl orthosilicato-kappao',di-s-butoxyaluminoxy-triethoxysilane,di-sec-butoxy triethoxysilyl oxy aluminum,bis sec-butoxy alumanyl triethyl silicate,aluminum,bis 2-butanolato triethyl orthosilicato-ko',diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane
16684138
di(butan-2-yloxy)alumanyl triethyl silicate
CCC(C)O[Al](OC(C)CC)O[Si](OCC)(OCC)OCC
68959-06-8
C14H33AlO6Si
352.479
MFCD00053771
MTHHTRVJDXYJHE-UHFFFAOYSA-N
di-sec-butoxyaluminoxytriethoxysilane,bis butan-2-olato triethyl orthosilicato-o' aluminium,aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o',aluminum, bis 2-butanolato triethyl orthosilicato-kappao',di-s-butoxyaluminoxy-triethoxysilane,di-sec-butoxy triethoxysilyl oxy aluminum,bis sec-butoxy alumanyl triethyl silicate,aluminum,bis 2-butanolato triethyl orthosilicato-ko',diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane
16684138
di(butan-2-yloxy)alumanyl triethyl silicate
CCC(C)O[Al](OC(C)CC)O[Si](OCC)(OCC)OCC
UN Number UN1993
Solubility Information Hydrolyzes with water.
Formula Weight 352.48
Physical Form Liquid
Density 1.00 g/mL
Sensitivity Moisture sensitive
Boiling Point >200°C
Color Colorless
Quantity 10g
Flash Point 46°C (115°F)
Chemical Name or Material Di-sec-butoxyaluminoxytriethoxysilane
Show More Show Less

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 273-392-6

TSCA : Yes

Recommended Storage : Ambient temperatures