Alfa Aesar™ Di-sec-butylamine, (+/-) + meso, 98%

Manufacturer: Alfa Aesar™ B25690-30

Quantity 250g

 View more versions of this product

Catalog No. AAB2569030

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 626-23-3
    Molecular Formula C8H19N
    Formula Weight 129.25
    MDL Number MFCD00009326
    UN Number UN2733
    Synonym Bis(2-methylpropyl)amine
    Chemical Name or Material Di-sec-butylamine, (+/-) + meso

    Specifications

    Formula Weight 129.25
    Melting Point −70°C
    TSCA TSCA
    Odor Amine-like
    MDL Number MFCD00009326
    UN Number UN2733
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H302-H312-H332
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 24°C (75°F)
    Boiling Point 134° to 136°C
    Quantity 250g
    Molecular Formula C8H19N
    CAS 626-23-3
    Synonym Bis(2-methylpropyl)amine
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Chemical Name or Material Di-sec-butylamine, (+/-) + meso
    Assay 98%

    GHS H Statement
    H225-H314-H302-H312-H332
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit