Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl dicarbonate, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
50g
100g
500g
Packaging:
Glass bottle
24424-99-5
C10H18O5
218.25
MFCD00008805
DYHSDKLCOJIUFX-UHFFFAOYSA-N
di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate
90495
CHEBI:48500
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
24424-99-5
C10H18O5
218.25
MFCD00008805
DYHSDKLCOJIUFX-UHFFFAOYSA-N
di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate
90495
CHEBI:48500
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
Packaging Glass bottle
Flash Point 37°C
Specific Gravity 1.02
Density 1.0200g/mL
Color White
Melting Point 22.0°C to 24.0°C
Quantity 10g
Refractive Index 1.4075 to 1.4095
Boiling Point 56.0°C to 57.0°C (0.5mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula O[CO2C(CH3)3]2
Fieser 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
Merck Index 15, 3038
Formula Weight 218.25
Physical Form Low Melting Crystalline Solid
Percent Purity 99%
Chemical Name or Material Di-tert-butyl dicarbonate, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti

GHS Signal Word: Danger

EINECSNumber : 246-240-1

RTECSNumber : HT0230000

TSCA : TSCA