Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl iminodicarboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
51779-32-9
C10H19NO4
217.27
XCAQIUOFDMREBA-UHFFFAOYSA-N
di-tert-butyl iminodicarboxylate,di-t-butyl iminodicarboxylate,di-tert-butyl imidodicarbonate,n-boc-tert-butylcarbamate,tert-butyl iminodicarboxylate,di-tert-butyl iminodicarbonate,di-tert-butyl-iminodicarboxylate,iminodicarboxylic acid di-tert-butyl ester,tert-butyl n-tert-butoxy carbonyl carbamate,di tert-butyl imidodicarbonate
279800
tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate
CC(C)(C)OC(=O)NC(=O)OC(C)(C)C
51779-32-9
C10H19NO4
217.27
XCAQIUOFDMREBA-UHFFFAOYSA-N
di-tert-butyl iminodicarboxylate,di-t-butyl iminodicarboxylate,di-tert-butyl imidodicarbonate,n-boc-tert-butylcarbamate,tert-butyl iminodicarboxylate,di-tert-butyl iminodicarbonate,di-tert-butyl-iminodicarboxylate,iminodicarboxylic acid di-tert-butyl ester,tert-butyl n-tert-butoxy carbonyl carbamate,di tert-butyl imidodicarbonate
279800
tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate
CC(C)(C)OC(=O)NC(=O)OC(C)(C)C
Quantity 25g
Infrared Spectrum Authentic
Color White to Beige
Melting Point 114.0°C to 117.0°C
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula [(CH3)3COCO]2NH
Fieser 01,210; 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268
Formula Weight 217.27
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Di-tert-butyl iminodicarboxylate
Show More Show Less

GHS Signal Word: Warning