Alfa Aesar™ Di-tert-butylphosphinoferrocene, 95%

Manufacturer: Alfa Aesar™ H2918303

Quantity 1g

 View more versions of this product

Catalog No. AAH2918303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 223655-16-1
    Molecular Formula C18H27FeP
    Formula Weight 330.23
    MDL Number MFCD06740718
    Chemical Name or Material Di-tert-butylphosphinoferrocene

    Specifications

    Formula Weight 330.23
    Molecular Formula C18H27FeP
    MDL Number MFCD06740718
    CAS 223655-16-1
    Melting Point 91° to 94°C
    Quantity 1g
    Chemical Name or Material Di-tert-butylphosphinoferrocene
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit