Alfa Aesar™ Di-tert-butyltin dichloride, 98%

Manufacturer: Alfa Aesar™ 3961902

Quantity 0.5g

 View more versions of this product

Catalog No. AA3961902

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19429-30-2
    Linear Formula [(CH3)3C]2SnCl2
    Formula Weight 303.83
    MDL Number MFCD00009854
    UN Number UN2928
    Chemical Name or Material Di-tert-butyltin dichloride

    Specifications

    Formula Weight 303.83
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White
    Linear Formula [(CH3)3C]2SnCl2
    Quantity 0.5g
    UN Number UN2928
    Flash Point >110° (230°F)
    Melting Point 42° to 43°C
    Physical Form Crystalline
    MDL Number MFCD00009854
    Health Hazard 1 Danger
    Odor Acrid
    Sensitivity Moisture Sensitive
    Boiling Point 66°C (3mmHg)
    CAS 19429-30-2
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Chemical Name or Material Di-tert-butyltin dichloride
    Assay 98%

    GHS H Statement
    H301-H311-H330-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit